Merck
  • Methicillin-resistant Staphylococcus saprophyticus isolates carrying staphylococcal cassette chromosome mec have emerged in urogenital tract infections.
Methicillin-resistant Staphylococcus saprophyticus isolates carrying staphylococcal cassette chromosome mec have emerged in urogenital tract infections.
Antimicrobial agents and chemotherapy (2008-03-26)
Masato Higashide, Makoto Kuroda, Carlos Takashi Neves Omura, Miyuki Kumano, Saburo Ohkawa, Sadahiro Ichimura, Toshiko Ohta
ZUSAMMENFASSUNG
Staphylococcus saprophyticus is a uropathogenic bacterium that causes acute uncomplicated urinary tract infections, particularly in female outpatients. We investigated the dissemination and antimicrobial susceptibilities of 101 S. saprophyticus isolates from the genitourinary tracts of patients in Japan. Eight of these isolates were mecA positive and showed beta-lactam resistance. Pulsed-field gel electrophoresis showed that only some isolates were isogenic, indicating that the mecA gene was apparently acquired independently by mecA-positive isolates through staphylococcal cassette chromosome mec (SCCmec). Type determination of SCCmec by multiplex PCR showed a nontypeable element in the eight mecA-positive isolates. Sequence analysis of the entire SCCmec element from a prototype S. saprophyticus strain revealed that it was nontypeable with the current SCCmec classification due to the novel composition of the class A mec gene complex (IS431-mecA-mecR1-mecI genes) and the ccrA1/ccrB3 gene complex. Intriguingly, the attachment sites of SCCmec are similar to those of type I SCCmec in S. aureus NCTC 10442. Furthermore, the genes around the mec gene complex are similar to those of type II/III SCCmec in S. aureus, while those around the ccr gene complex are similar to those of SCC15305RM found in S. saprophyticus ATCC 15305. In comparison with known SCCmec elements, this S. saprophyticus SCCmec is a novel type.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.