Merck
  • Design, synthesis and in vivo study of novel pyrrolidine-based 11β-HSD1 inhibitors for age-related cognitive dysfunction.
Design, synthesis and in vivo study of novel pyrrolidine-based 11β-HSD1 inhibitors for age-related cognitive dysfunction.
European journal of medicinal chemistry (2017-08-19)
Rosana Leiva, Christian Griñan-Ferré, Constantí Seira, Elena Valverde, Andrew McBride, Margaret Binnie, Belén Pérez, F Javier Luque, Mercè Pallàs, Axel Bidon-Chanal, Scott P Webster, Santiago Vázquez
ZUSAMMENFASSUNG
Recent findings suggest that treatment with 11β-HSD1 inhibitors provides a novel approach to deal with age-related cognitive dysfunctions, including Alzheimer's disease. In this work we report potent 11β-HSD1 inhibitors featuring unexplored pyrrolidine-based polycyclic substituents. A selected candidate administered to 12-month-old SAMP8 mice for four weeks prevented memory deficits and displayed a neuroprotective action. This is the first time that 11β-HSD1 inhibitors have been studied in this broadly-used mouse model of accelerated senescence and late-onset Alzheimer's disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-KDR/Flk-1/VEGFR2 Antibody, clone CH-11, clone CH-11, Upstate®, from mouse