Merck
  • The RNA binding protein EWS is broadly involved in the regulation of pri-miRNA processing in mammalian cells.
The RNA binding protein EWS is broadly involved in the regulation of pri-miRNA processing in mammalian cells.
Nucleic acids research (2018-07-28)
Huiwu Ouyang, Kai Zhang, Kristi Fox-Walsh, Yang Yang, Chen Zhang, Jie Huang, Hairi Li, Yu Zhou, Xiang-Dong Fu
ZUSAMMENFASSUNG
The Ewing Sarcoma protein (EWS) is a multifaceted RNA binding protein (RBP) with established roles in transcription, pre-mRNA processing and DNA damage response. By generating high quality EWS-RNA interactome, we uncovered its specific and prevalent interaction with a large subset of primary microRNAs (pri-miRNAs) in mammalian cells. Knockdown of EWS reduced, whereas overexpression enhanced, the expression of its target miRNAs. Biochemical analysis revealed that multiple elements in target pri-miRNAs, including the sequences flanking the stem-loop region, contributed to high affinity EWS binding and sequence swap experiments between target and non-target demonstrated that the flanking sequences provided the specificity for enhanced pri-miRNA processing by the Microprocessor Drosha/DGCR8. Interestingly, while repressing Drosha expression, as reported earlier, we found that EWS was able to enhance the recruitment of Drosha to chromatin. Together, these findings suggest that EWS may positively and negatively regulate miRNA biogenesis via distinct mechanisms, thus providing a new foundation to understand the function of EWS in development and disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Roche
DIG Northern-Starterkit, suitable for Northern blotting, sufficient for 10 labeling reactions