Merck
  • Candida albicans is able to use M cells as a portal of entry across the intestinal barrier in vitro.
Candida albicans is able to use M cells as a portal of entry across the intestinal barrier in vitro.
Cellular microbiology (2015-08-06)
Sandrine Albac, Antonin Schmitz, Carolina Lopez-Alayon, Christophe d'Enfert, Marc Sautour, Amandine Ducreux, Catherine Labruère-Chazal, Michael Laue, Gudrun Holland, Alain Bonnin, Frederic Dalle
ZUSAMMENFASSUNG
Candida albicans is the most frequent yeast responsible for systemic infections in humans. These infections mainly originate from the gastrointestinal tract where C. albicans can invade the gut epithelial barrier to gain access to the bloodstream. Along the gut, pathogens can use Microfold (M) cells as a portal of entry to cross the epithelial barrier. M cells are specialized cells mainly located in the follicule-associated epithelium of Peyer patches. In this study, we used scanning electron and fluorescence microscopy, adhesion and invasion assays and fungal mutants to investigate the interactions of C. albicans with M cells obtained in an established in vitro model whereby enterocyte-like Caco-2 cells co-cultured with the Raji B cell line undergo a phenotypic switch to morphologically and functionally resembling M cells. Our data demonstrate that C. albicans co-localizes with and invades preferentially M cells, providing evidence that the fungus can use M cells as a portal of entry into the intestinal barrier. In addition to active penetration, F-actin dependent endocytosis contributes to internalization of the fungus into M cells through a mechanism involving hypha-associated invasins including Ssa1 and Als3.