Merck
  • Application of a proteolysis/mass spectrometry method for investigating the effects of inhibitors on hydroxylase structure.
Application of a proteolysis/mass spectrometry method for investigating the effects of inhibitors on hydroxylase structure.
Journal of medicinal chemistry (2009-04-15)
Christopher J Stubbs, Christoph Loenarz, Jasmin Mecinović, Kar Kheng Yeoh, Nicola Hindley, Benoît M Liénard, Frank Sobott, Christopher J Schofield, Emily Flashman
ZUSAMMENFASSUNG
Limited proteolysis coupled to matrix-assisted laser desorption/ionization (MALDI) mass spectrometric analyses can be used to screen for compounds that alter protein structure by monitoring stabilizing/destabilizing effects with respect to the rate and nature of proteolysis. When applied to prolyl hydroxylase 2, a key enzyme involved in human oxygen sensing, the method efficiently revealed differential effects on proteolytic stability for structurally similar compounds and for different substrates.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fumarsäure, ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
α-Ketoglutarsäure, 99.0-101.0% (T)
Sigma-Aldrich
α-Ketoglutarsäure, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Bernsteinsäure, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
α-Ketoglutarsäure, ≥98.5% (NaOH, titration)
Sigma-Aldrich
Bernsteinsäure, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Bernsteinsäure, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Bernsteinsäure, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
Fumarsäure, FCC, FG