Merck
  • IL-6/Stat3-Dependent Induction of a Distinct, Obesity-Associated NK Cell Subpopulation Deteriorates Energy and Glucose Homeostasis.
IL-6/Stat3-Dependent Induction of a Distinct, Obesity-Associated NK Cell Subpopulation Deteriorates Energy and Glucose Homeostasis.
Cell metabolism (2017-07-07)
Sebastian Theurich, Eva Tsaousidou, Ruth Hanssen, Adelheid M Lempradl, Jan Mauer, Katharina Timper, Katharina Schilbach, Kat Folz-Donahue, Christian Heilinger, Veronika Sexl, John Andrew Pospisilik, F Thomas Wunderlich, Jens C Brüning
ZUSAMMENFASSUNG
Natural killer (NK) cells contribute to the development of obesity-associated insulin resistance. We demonstrate that in mice obesity promotes expansion of a distinct, interleukin-6 receptor (IL6R)a-expressing NK subpopulation, which also expresses a number of other myeloid lineage genes such as the colony-stimulating factor 1 receptor (Csf1r). Selective ablation of this Csf1r-expressing NK cell population prevents obesity and insulin resistance. Moreover, conditional inactivation of IL6Ra or Stat3 in NK cells limits obesity-associated formation of these myeloid signature NK cells, protecting from obesity, insulin resistance, and obesity-associated inflammation. Also in humans IL6Ra+ NK cells increase in obesity and correlate with markers of systemic low-grade inflammation, and their gene expression profile overlaps with characteristic gene sets of NK cells in obese mice. Collectively, we demonstrate that obesity-associated inflammation and metabolic disturbances depend on interleukin-6/Stat3-dependent formation of a distinct NK population, which may provide a target for the treatment of obesity, metaflammation-associated pathologies, and diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Calnexin-Antikörper vom Kaninchen mit C-Terminus (575–593) pAb, liquid, Calbiochem®