Merck
11-Phenyl-[b,e]-dibenzazepine compounds: novel antitumor agents.
Bioorganic & medicinal chemistry letters (2008-11-26)
Raed A Al-Qawasmeh, Yoon Lee, Ming-Yu Cao, Xiaoping Gu, Stéphane Viau, Jeff Lightfoot, Jim A Wright, Aiping H Young
ZUSAMMENFASSUNG
A series of 11-phenyl-[b,e]-dibenzazepine compounds were synthesized and shown to be inhibitors of tumor cell proliferation with IC(50) values ranging from submicromolar to micromolar concentrations. Flow cytometric analyses of several active compounds demonstrated inhibition of cell cycle progression at the G(0)-G(1) phase transition resulting in G(0)-G(1) arrest.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Clotrimazol