Merck
Full Issue PDF.
JACC. Basic to translational science (2020-10-06)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®