Merck
  • Biological Evaluation of Newly Synthesized Biaryl Guanidine Derivatives to Arrest β-Secretase Enzymatic Activity Involved in Alzheimer's Disease.
Biological Evaluation of Newly Synthesized Biaryl Guanidine Derivatives to Arrest β-Secretase Enzymatic Activity Involved in Alzheimer's Disease.
BioMed research international (2020-05-29)
Sayyad Ali, Muhammad Hassham Hassan Bin Asad, Fahad Khan, Ghulam Murtaza, Albert A Rizvanov, Jamshed Iqbal, Borhan Babak, Izhar Hussain
ZUSAMMENFASSUNG
Proteases BACE1 (β-secretases) enzymes have been recognized as a promising target associated with Alzheimer's disease (AD). This study was carried out on the principles of molecular docking, chemical synthesis, and enzymatic inhibition of BACE1 enzymes via biaryl guanidine-based ligands. Based on virtual screening, thirteen different compounds were synthesized and subsequently evaluated via in vitro and in vivo studies. Among them, 1,3-bis(5,6-difluoropyridin-3-yl)guanidine (compound (9)) was found the most potent (IC50 = 97 ± 0.91 nM) and active to arrest (99%) β-secretase enzymes (FRET assay). Furthermore, it was found to improve the novel object recognition test and Morris water maze test significantly (p < 0.05). Improved pharmacokinetic parameters, viz., Log P o/w (1.76), Log S (-2.73), and better penetration to the brain (BBB permeation) with zero Lipinski violation, made it possible to hit the BACE1 as a potential therapeutic source for AD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Secretase (BACE1) Activity Detection Kit (Fluorescent), 1 kit sufficient for 250 reactions