- Complete Genome Sequence of Staphylococcus carnosus LTH 3730.
Complete Genome Sequence of Staphylococcus carnosus LTH 3730.
Genome announcements (2016-10-01)
Anne Müller, Jochen Klumpp, Herbert Schmidt, Agnes Weiss
PMID27688338
ZUSAMMENFASSUNG
Specific strains of the apathogenic coagulase-negative species Staphylococcus carnosus are frequently used as meat starter cultures, as they contribute to color formation and the production of aroma compounds. Here, we report the complete genome sequence of S. carnosus LTH 3730, a strain isolated from a fermented fish product.