Merck
  • Nanoparticles (NPs)-Meditated LncRNA AFAP1-AS1 Silencing to Block Wnt/β-Catenin Signaling Pathway for Synergistic Reversal of Radioresistance and Effective Cancer Radiotherapy.
Nanoparticles (NPs)-Meditated LncRNA AFAP1-AS1 Silencing to Block Wnt/β-Catenin Signaling Pathway for Synergistic Reversal of Radioresistance and Effective Cancer Radiotherapy.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2020-10-02)
Zhuofei Bi, Qingjian Li, Xiaoxiao Dinglin, Ying Xu, Kaiyun You, Huangming Hong, Qian Hu, Wei Zhang, Chenchen Li, Yujie Tan, Ning Xie, Wei Ren, Chuping Li, Yimin Liu, Hai Hu, Xiaoding Xu, Herui Yao
ZUSAMMENFASSUNG
Resistance to radiotherapy is frequently encountered in clinic, leading to poor prognosis of cancer patients. Long noncoding RNAs (lncRNAs) play important roles in the development of radioresistance due to their functions in regulating the expression of target genes at both transcriptional and posttranscriptional levels. Exploring key lncRNAs and elucidating the mechanisms contributing to radioresistance are crucial for the development of effective strategies to reverse radioresistance, which however remains challenging. Here, actin filament-associated protein 1 antisense RNA1 (lncAFAP1-AS1) is identified as a key factor in inducing radioresistance of triple-negative breast cancer (TNBC) via activating the Wnt/β-catenin signaling pathway. Considering the generation of a high concentration of reduction agent glutathione (GSH) under radiation, a reduction-responsive nanoparticle (NP) platform is engineered for effective lncAFAP1-AS1 siRNA (siAFAP1-AS1) delivery. Systemic delivery of siAFAP1-AS1 with the reduction-responsive NPs can synergistically reverse radioresistance by silencing lncAFAP1-AS1 expression and scavenging intracellular GSH, leading to a dramatically enhanced radiotherapy effect in both xenograft and metastatic TNBC tumor models. The findings indicate that lncAFAP1-AS1 can be used to predict the outcome of TNBC radiotherapy and combination of systemic siAFAP1-AS1 delivery with radiotherapy can be applied for the treatment of recurrent TNBC patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Magna RIP® RNA-Bindungsprotein Immunpräzipitations-Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.