Merck
  • Synthetic staurosporines via a ring closing metathesis strategy as potent JAK3 inhibitors and modulators of allergic responses.
Synthetic staurosporines via a ring closing metathesis strategy as potent JAK3 inhibitors and modulators of allergic responses.
Bioorganic & medicinal chemistry letters (2009-05-12)
Lawrence J Wilson, Ravi Malaviya, Cangming Yang, Rochelle Argentieri, Bingbing Wang, Xin Chen, William V Murray, Druie Cavender
ZUSAMMENFASSUNG
The synthesis and biological evaluation of JAK3 based staurosporine compounds is described. The compounds are constructed completely de novo, and a ring closing metathesis strategy is used to assemble the sugar mimetic portion. These analogs show potent JAK3 activity against isolated enzyme and in T-cells. One analog (32) showed unique biological effects during in vitro and in vivo tests including inhibition of STAT5 phosphorylation, blockade of mast cell responses, and reduction of JAK3 based effects in mice models of allergic disease.