Merck
Human Cdc45 is a proliferation-associated antigen.
The FEBS journal (2007-07-05)
S Pollok, C Bauerschmidt, J Sänger, H-P Nasheuer, F Grosse
ZUSAMMENFASSUNG
Cell division cycle protein 45 (Cdc45) plays a critical role in DNA replication to ensure that chromosomal DNA is replicated only once per cell cycle. We analysed the expression of human Cdc45 in proliferating and nonproliferating cells. Our findings show that Cdc45 protein is absent from long-term quiescent, terminally differentiated and senescent human cells, although it is present throughout the cell cycle of proliferating cells. Moreover, Cdc45 is much less abundant than the minichromosome maintenance (Mcm) proteins in human cells, supporting the concept that origin binding of Cdc45 is rate limiting for replication initiation. We also show that the Cdc45 protein level is consistently higher in human cancer-derived cells compared with primary human cells. Consequently, tumour tissue is preferentially stained using Cdc45-specific antibodies. Thus, Cdc45 expression is tightly associated with proliferating cell populations and Cdc45 seems to be a promising candidate for a novel proliferation marker in cancer cell biology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Monoklonales Anti-β-Tubulin-I in Maus hergestellte Antikörper, clone SAP.4G5, ascites fluid