Merck
  • Transcriptional orchestration of mitochondrial homeostasis in a cellular model of PGC-1-related coactivator-dependent thyroid tumor.
Transcriptional orchestration of mitochondrial homeostasis in a cellular model of PGC-1-related coactivator-dependent thyroid tumor.
Oncotarget (2018-04-18)
Solenne Dumont, Soazig Le Pennec, Audrey Donnart, Raluca Teusan, Marja Steenman, Catherine Chevalier, Rémi Houlgatte, Frédérique Savagner
ZUSAMMENFASSUNG
The PGC-1 (Peroxisome proliferator-activated receptor Gamma Coactivator-1) family of coactivators (PGC-1α, PGC-1β, and PRC) plays a central role in the transcriptional control of mitochondrial biogenesis and oxidative phosphorylation (OXPHOS) processes. These coactivators integrate mitochondrial energy production into cell metabolism using complementary pathways. The XTC.UC1 cell line is a mitochondria-rich model of thyroid tumors whose biogenesis is almost exclusively dependent on PRC. Here we aim to propose an integrative view of the cellular pathways regulated by PRC through integration of cDNA and miRNA microarray data and chromatin immunoprecipitation results obtained from XTC.UC1 cells invalidated for PRC. This study showes that PRC induces a complex network of cellular functions interacting with at least one to five of the studied transcription factors (Estrogen Related Receptor alpha, ERR1; Nuclear-Respiratory Factors, NRF1 and NRF2; cAMP Response Element Binding, CREB; and Ying Yang, YY1). Our data confirm that ERR1 is a key partner of PRC in the regulation of mitochondrial functions and suggest a potential role of this complex in RNA processing. PRC is also involved in transcriptional regulatory complexes targeting 12 miRNAs, five of which are involved in the control of the OXPHOS process. Our findings demonstrate that the PRC coactivator can act in complex with several transcription factors and regulate miRNA expression to control the fine regulation of main metabolic functions in the cell. Therefore, in PGC-1α/β-associated pathologies, PRC, as a metabolic sensor, may ensure mitochondrial homeostasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenomePlex® GGA Kit zur Gesamtgenom-Amplifikation, Optimized kit with enzyme for amplifying a variety of DNA including FFPE tissue