- Synthesis and antifungal activity of benzo[d]oxazole-4,7-diones.
Synthesis and antifungal activity of benzo[d]oxazole-4,7-diones.
Bioorganic & medicinal chemistry letters (2009-09-08)
Chung-Kyu Ryu, Ra-Young Lee, Na Young Kim, Yang Hui Kim, Ae Li Song
PMID19733068
ZUSAMMENFASSUNG
Benzo[d]oxazole-4,7-diones were synthesized and tested for in vitro antifungal activity against fungi. Among them tested, many compounds showed good antifungal activity. The results suggest that benzo[d]oxazole-4,7-diones would be potent antifungal agents.
MATERIALIEN