Merck
Metabolic engineering of Corynebacterium glutamicum for L-arginine production.
Nature communications (2014-08-06)
Seok Hyun Park, Hyun Uk Kim, Tae Yong Kim, Jun Seok Park, Suok-Su Kim, Sang Yup Lee
ZUSAMMENFASSUNG
L-arginine is an important amino acid for diverse industrial and health product applications. Here we report the development of metabolically engineered Corynebacterium glutamicum ATCC 21831 for the production of L-arginine. Random mutagenesis is first performed to increase the tolerance of C. glutamicum to L-arginine analogues, followed by systems metabolic engineering for further strain improvement, involving removal of regulatory repressors of arginine operon, optimization of NADPH level, disruption of L-glutamate exporter to increase L-arginine precursor and flux optimization of rate-limiting L-arginine biosynthetic reactions. Fed-batch fermentation of the final strain in 5 l and large-scale 1,500 l bioreactors allows production of 92.5 and 81.2 g l(-1) of L-arginine with the yields of 0.40 and 0.35 g L-arginine per gram carbon source (glucose plus sucrose), respectively. The systems metabolic engineering strategy described here will be useful for engineering Corynebacteria strains for the industrial production of L-arginine and related products.