Merck
Liver kinase B1 inhibits smooth muscle calcification via high mobility group box 1.
Redox biology (2020-12-19)
Tianran Zhang, Hongxuan Li, Changhan Ouyang, Guangqing Cao, Jiangang Gao, Jiliang Wu, Jianmin Yang, Nengwang Yu, Qing Min, Cheng Zhang, Wencheng Zhang
ZUSAMMENFASSUNG
Vascular calcification is a common pathological feature of atherosclerosis, chronic kidney disease, vascular injury, and aging. Liver kinase B1 (LKB1) plays pivotal roles in cellular processes such as apoptosis, metabolism, and cell cycle regulation. In addition, growing evidence has indicated that LKB1 functions as a tumor suppressor gene. However, its role in vascular calcification has not been reported. LKB1flox/flox mice were hybridized with SM22-CreERT2 transgenic mice and adult mice received tamoxifen to obtain smooth muscle-specific LKB1-knockout (LKB1SMKO) mice. LKB1 expression was decreased under calcifying conditions, and LKB1 overexpression had a protective effect on vascular calcification. However, high mobility group box 1 (HMGB1) overexpression partially counteracted the promotion of vascular calcification induced by LKB1 overexpression. Mechanically, LKB1 could bind to HMGB1 to promote HMGB1 degradation. Furthermore, LKB1SMKO mice showed intensified vascular calcification, which was alleviated by treatment with the HMGB1 inhibitor glycyrrhizic acid. Based on our results, LKB1 may inhibit vascular calcification via inhibiting HMGB1 expression.