Merck
  • Draft Genome Sequence of Muricauda sp. Strain K001 Isolated from a Marine Cyanobacterial Culture.
Draft Genome Sequence of Muricauda sp. Strain K001 Isolated from a Marine Cyanobacterial Culture.
Genome announcements (2018-06-02)
Carla S Vizzotto, Fabyano A C Lopes, Stefan J Green, Andrei S Steindorff, Juline M Walter, Fabiano L Thompson, Ricardo H Krüger
ZUSAMMENFASSUNG
We report the whole-genome sequence of Muricauda sp. strain K001 isolated from a marine cyanobacterial culture. This genome sequence will improve our understanding of the influence of heterotrophic bacteria on the physiology of cyanobacteria and may contribute to the development of new natural products.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenElute Bacterial Genomic DNA Kit, sufficient for 70 purifications