Merck
  • miR-212 downregulation contributes to the protective effect of exercise against non-alcoholic fatty liver via targeting FGF-21.
miR-212 downregulation contributes to the protective effect of exercise against non-alcoholic fatty liver via targeting FGF-21.
Journal of cellular and molecular medicine (2015-12-10)
Junjie Xiao, Yihua Bei, Jingqi Liu, Jasmina Dimitrova-Shumkovska, Dapeng Kuang, Qiulian Zhou, Jin Li, Yanning Yang, Yang Xiang, Fei Wang, Changqing Yang, Wenzhuo Yang
ZUSAMMENFASSUNG
Non-alcoholic fatty liver disease (NAFLD) is associated with obesity and lifestyle, while exercise is beneficial for NAFLD. Dysregulated microRNAs (miRs) control the pathogenesis of NAFLD. However, whether exercise could prevent NAFLD via targeting microRNA is unknown. In this study, normal or high-fat diet (HF) mice were either subjected to a 16-week running program or kept sedentary. Exercise attenuated liver steatosis in HF mice. MicroRNA array and qRT-PCR demonstrated that miR-212 was overexpressed in HF liver, while reduced by exercise. Next, we investigated the role of miR-212 in lipogenesis using HepG2 cells with/without long-chain fatty acid treatment (± FFA). FFA increased miR-212 in HepG2 cells. Moreover, miR-212 promoted lipogenesis in HepG2 cells (± FFA). Fibroblast growth factor (FGF)-21, a key regulator for lipid metabolism, was negatively regulated by miR-212 at protein level in HepG2 cells. Meanwhile, FFA downregulated FGF-21 both at mRNA and protein levels in HepG2 cells. Also, FGF-21 protein level was reduced in HF liver, while reversed by exercise in vivo. Furthermore, siRNA-FGF-21 abolished the lipogenesis-reducing effect of miR-212 inhibitor in HepG2 cells (± FFA), validating FGF-21 as a target gene of miR-212. These data link the benefit of exercise and miR-212 downregulation in preventing NAFLD via targeting FGF-21.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-FGF21 Mouse mAb (2F11), liquid, clone 2F11, Calbiochem®