- Probing of the cis-5-phenyl proline scaffold as a platform for the synthesis of mechanism-based inhibitors of the Staphylococcus aureus sortase SrtA isoform.
Probing of the cis-5-phenyl proline scaffold as a platform for the synthesis of mechanism-based inhibitors of the Staphylococcus aureus sortase SrtA isoform.
Bioorganic & medicinal chemistry (2009-03-10)
Konstantin V Kudryavtsev, Matthew L Bentley, Dewey G McCafferty
PMID19269184
ZUSAMMENFASSUNG
cis-5-Phenyl prolinates with electrophilic substituents at the fourth position of a pyrrolidine ring were synthesized by 1,3-dipolar cycloaddition of arylimino esters with divinyl sulfone and acrylonitrile. 4-Vinylsulfonyl 5-phenyl prolinates inhibit Staphylococcus aureus sortase SrtA irreversibly by modification of the enzyme Cys184 and could be used as hits for the development of antibacterials and antivirulence agents.