Merck
  • Drug-tunable multidimensional synthetic gene control using inducible degron-tagged dCas9 effectors.
Drug-tunable multidimensional synthetic gene control using inducible degron-tagged dCas9 effectors.
Nature communications (2017-11-01)
Dirk A Kleinjan, Caroline Wardrope, Si Nga Sou, Susan J Rosser
ZUSAMMENFASSUNG
The nuclease-deactivated variant of CRISPR-Cas9 proteins (dCas9) fused to heterologous transactivation domains can act as a potent guide RNA sequence-directed inducer or repressor of gene expression in mammalian cells. In such a system the long-term presence of a stable dCas9 effector can be a draw-back precluding the ability to switch rapidly between repressed and activated target gene expression states, imposing a static environment on the synthetic regulatory circuits in the cell. To address this issue we have generated a toolkit of conditionally degradable or stabilisable orthologous dCas9 or Cpf1 effector proteins, thus opening options for multidimensional control of functional activities through combinations of orthogonal, drug-tunable artificial transcription factors.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LookOut® Mycoplasma PCR-Detektions-Kit, Optimized for use with JumpStart Taq DNA Polymerase, D9307.
Sigma-Aldrich
Monoklonaler ANTI-HA-Antikörper in Maus hergestellte Antikörper, clone HA-7, ascites fluid
Sigma-Aldrich
Anti-Caspase 9 antibody,Mouse monoclonal, clone CAS9, purified from hybridoma cell culture