Merck
p-Terphenyls from the fruiting bodies of Paxillus curtisii and their antioxidant properties.
Bioorganic & medicinal chemistry (2009-05-20)
In-Kyoung Lee, Jin-Young Jung, Young-Sook Kim, Man Hee Rhee, Bong-Sik Yun
ZUSAMMENFASSUNG
New p-terphenyls, curtisians R (1)-V (5), have been isolated together with known compounds, curtisians E, I-P, and kynapcin-12, from the methanolic extract of the fruiting bodies of Paxillus curtisii (Paxillaceace). Their structures were established by various spectroscopic analyses including 1D- and 2D-NMR experiments, as well as high-resolution FAB-mass analysis. They exhibited significant protective effects against oxidative damage of supercoiled DNA and 2-deoxyribose by hydroxyl radicals generated from the Fenton reaction.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)