Merck
  • Combined local ischemic postconditioning and remote perconditioning recapitulate cardioprotective effects of local ischemic preconditioning.
Combined local ischemic postconditioning and remote perconditioning recapitulate cardioprotective effects of local ischemic preconditioning.
American journal of physiology. Heart and circulatory physiology (2010-03-09)
Ping Xin, Wei Zhu, Jing Li, Shixin Ma, Lixing Wang, Mingya Liu, Jingbo Li, Meng Wei, Andrew N Redington
ZUSAMMENFASSUNG
Ischemic postconditioning (PostC) and perconditioning (PerC) provide practical methods for protecting the heart against ischemia-reperfusion (I/R) injury, but their combined effects have not been studied in detail. Using an in vivo rat I/R model, we tested 1) whether additive effects were produced when local PostC was preceded by varying doses of remote PerC, and whether the optimal PostC+PerC regime is additive to local ischemic preconditioning (IPC), and 2) how combined PostC+PerC alters the activity of the reperfusion injury salvage kinase pathway. The optimal combination of PerC and PostC therapy was produced by PerC delivered with four cycles of 5 min of limb ischemia followed by 5-min reperfusion. This resulted in lower infarct size (22.56 +/- 4.45%) compared with rats with PostC alone (29.39 +/- 3.66%) and PerC alone (33.49 +/- 5.81%) and complementary differences in the generation of reactive oxygen species and apoptotic signaling. However, this optimal combination of PostC+PerC resulted in protection similar to local IPC alone (18.8 +/- 2.54%, P = 0.13), and when added to IPC there was no additional protection (19.62 +/- 2.89%, P = 0.675). Akt and ERK1/2 phosphorylation was induced by PostC and PerC and maximally by combined PostC+PerC treatment, and protection was abolished by phosphatidylinositol 3-kinase or ERK1/2 inhibitors. This study shows that neither PostC nor a maximized "dose" of PerC leads to optimal kinase signaling or cardioprotection compared with IPC alone. However, combined PostC+PerC may result in complementary effects on kinase signaling to recapitulate the effects of local IPC. Finally, combined PostC+PerC is not additive to IPC, suggesting that each works via a common pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
Lipid Hydroperoxide (LPO) Assay Kit