- Draft Genome Sequence of Pseudomonas aeruginosa SG17M, an Environmental Isolate Belonging to Clone C, Prevalent in Patients and Aquatic Habitats.
Draft Genome Sequence of Pseudomonas aeruginosa SG17M, an Environmental Isolate Belonging to Clone C, Prevalent in Patients and Aquatic Habitats.
Genome announcements (2014-03-22)
Changhan Lee, Verena Peters, Ojar Melefors, Ute Römling
PMID24652978
ZUSAMMENFASSUNG
Pseudomonas aeruginosa SG17M is an environmental isolate recovered from river water in the city of Mulheim, Germany. SG17M belongs to clone C, which is distributed worldwide. This is the first clone C strain whose genome sequence has been determined.