Merck
Prospecting Biomarkers for Diagnostic and Therapeutic Approaches in Pythiosis.
Journal of fungi (Basel, Switzerland) (2021-06-03)
Jéssica Luana Chechi, Tiwa Rotchanapreeda, Giselle Souza da Paz, Ana Carolina Prado, Alana Lucena Oliveira, José Cavalcante Souza Vieira, Marília Afonso Rabelo Buzalaf, Anderson Messias Rodrigues, Lucilene Delazari Dos Santos, Theerapong Krajaejun, Sandra de Moraes Gimenes Bosco
ZUSAMMENFASSUNG
Pythiosis, whose etiological agent is the oomycete Pythium insidiosum, is a life-threatening disease that occurs mainly in tropical and subtropical countries, affecting several animal species. It is frequently found in horses in Brazil and humans in Thailand. The disease is difficult to diagnose because the pathogen's hyphae are often misdiagnosed as mucoromycete fungi in histological sections. Additionally, there is no specific antigen to use for rapid diagnosis, the availability of which could improve the prognosis in different animal species. In this scenario, we investigated which P. insidiosum antigens are recognized by circulating antibodies in horses and humans with pythiosis from Brazil and Thailand, respectively, using 2D immunoblotting followed by mass spectrometry for the identification of antigens. We identified 23 protein spots, 14 recognized by pooled serum from horses and humans. Seven antigens were commonly recognized by both species, such as the heat-shock cognate 70 KDa protein, the heat-shock 70 KDa protein, glucan 1,3-beta-glucosidase, fructose-bisphosphate aldolase, serine/threonine-protein phosphatase, aconitate hydratase, and 14-3-3 protein epsilon. These results demonstrate that there are common antigens recognized by the immune responses of horses and humans, and these antigens may be studied as biomarkers for improving diagnosis and treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Horse IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution