Merck
Computational modeling of novel inhibitors targeting the Akt pleckstrin homology domain.
Bioorganic & medicinal chemistry (2009-09-08)
Lei Du-Cuny, Zuohe Song, Sylvestor Moses, Garth Powis, Eugene A Mash, Emmanuelle J Meuillet, Shuxing Zhang
ZUSAMMENFASSUNG
Computational modeling continues to play an important role in novel therapeutics discovery and development. In this study, we have investigated the use of in silico approaches to develop inhibitors of the pleckstrin homology (PH) domain of AKT (protein kinase B). Various docking/scoring schemes have been evaluated, and the best combination was selected to study the system. Using this strategy, two hits were identified and their binding behaviors were investigated. Robust and predictive QSAR models were built using the k nearest neighbor (kNN) method to study their cellular permeability. Based on our in silico results, long flexible aliphatic tails were proposed to improve the Caco-2 penetration without affecting the binding mode. The modifications enhanced the AKT inhibitory activity of the compounds in cell-based assays, and increased their activity as in vivo antitumor testing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Koffein, powder, ReagentPlus®
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Hydrocortison -Lösung, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
β-Estradiol, ≥98%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
(−)-Nikotin, ≥99% (GC), liquid
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Salicylsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Millipore
Saccharose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Theophyllin, anhydrous, ≥99%, powder
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Acetylsalicylsäure, ≥99.0%
Sigma-Aldrich
(−)-Epinephrin
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture