Merck
  • New insights into the antioxidant activity of hydroxycinnamic acids: Synthesis and physicochemical characterization of novel halogenated derivatives.
New insights into the antioxidant activity of hydroxycinnamic acids: Synthesis and physicochemical characterization of novel halogenated derivatives.
European journal of medicinal chemistry (2008-12-09)
Alexandra Gaspar, E Manuela Garrido, Mário Esteves, Elias Quezada, Nuno Milhazes, Jorge Garrido, Fernanda Borges
ZUSAMMENFASSUNG
An interdisciplinary research project was developed combining the synthesis of a series of hydroxycinnamic acid derivatives and the evaluation of their physicochemical parameters (namely redox potentials and partition coefficients), along with the corresponding antioxidant activity. A structure-property-activity relationship (SPAR) approach was then applied aiming at establishing a putative relation between the physicochemical parameters of the compounds under study and their antioxidant activity. The results gathered allow concluding that the redox potentials could contribute to the understanding of the antioxidant activity and that the presence of an electron withdrawing group (EWG) of halogen type, namely a bromo atom, in an ortho position to a phenolic group of the cinnamic scaffold does not influence the antioxidant activity. On the other hand after the introduction of this type of substituent a significant increase on the lipophilicity of cinnamic derivatives was observed, which is a feature of extreme importance in the development of novel lipophilic antioxidants. The SPAR results revealed a relation between the redox potentials and the antioxidant activity of hydroxycinnamic acids and derivatives. The data obtained operate as a positive reinforce of the tendency to use redox properties as a guideline of the rational design of this type of compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Ferulasäure, 99%
Sigma-Aldrich
Sinapinsäure, ≥98%, powder
Sigma-Aldrich
trans-Ferulasäure, ≥99%
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
trans-Ferulasäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Kaffeesäure, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Sinapinsäure, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
trans-Ferulasäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland