Merck
  • Inhibition of eNOS by L-NAME resulting in rat hind limb developmental defects through PFKFB3 mediated angiogenetic pathway.
Inhibition of eNOS by L-NAME resulting in rat hind limb developmental defects through PFKFB3 mediated angiogenetic pathway.
Scientific reports (2020-10-09)
Ziqiang Wu, Huan Yao, Huan Xu, Yang Wang, Wangming Hu, Guanhua Lou, Lingling Zhang, Cong Huang, Cen Jiang, Shiyi Zhou, Yaping Shi, Xiongbing Chen, Lan Yang, Yiming Xu, Yong Wang
ZUSAMMENFASSUNG
L-arginine/NOS/NO signaling pathway plays a critical role in controlling variety of vascular diseases. However, whether NOS inhibition by L-NAME suppresses late embryonic development is undefined. The aim of this study is to determine whether NOS inhibition by L-NAME is critical for late embryonic rat hind limb development. The pregnant rat at E13.5 administrated L-NAME by consecutive intraperitoneal injection. The embryos been harvested from E16.5 to E 20.5. Hematoxylin and Eosin Staining, Immunofluorescence and Immunohistochemistry performed to determine hind limb Vasculogenesis, HUVEC culture, Adenoviral PFKFB3 infection, Real time PCR and western blot were performed to determine whether L-arginine/NOS/NO pathway controlling late embryonic hind limb development through PFKFB3 mediated angiogenetic pathway. NOS inhibition by L-NAME resulting in late embryonic hind limb developmental defects characterized by severe hemorrhage. The in vivo studies showed that NOS inhibition strongly suppressed hind limb angiogenetic remodeling by impairing differentiation of endothelial cells and smooth muscle cells, and extracellular matrix synthesis. For underlie mechanism, our studies indicated that L-NAME treatment dramatically suppresses PFKFB3 expression in hematopoietic progenitor cells, tubulogenetic endothelial cells and smooth muscle cells. Knockdown of PFKFB3 dramatically inhibits the expression of angiogenetic genes, as well as tubulogenesis and extracellular matrix related genes. Taken together, our data in this study demonstrated that L-arginine-eNOS-NO pathway is important for rat hind limb development during late embryonic stage. This could be both a useful animal model and a promising therapeutic treatment for defects of late embryonic developmental hind limbs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nω-Nitro-L-Arginin-Methylester -hydrochlorid, ≥97% (TLC), powder