Merck
  • High-mobility group nucleosomal binding domain 2 protects against microcephaly by maintaining global chromatin accessibility during corticogenesis.
High-mobility group nucleosomal binding domain 2 protects against microcephaly by maintaining global chromatin accessibility during corticogenesis.
The Journal of biological chemistry (2019-11-09)
Xue-Ling Gao, Wen-Jia Tian, Bofeng Liu, Jingyi Wu, Wei Xie, Qin Shen
ZUSAMMENFASSUNG
The surface area of the human cerebral cortex undergoes dramatic expansion during late fetal development, leading to cortical folding, an evolutionary feature not present in rodents. Microcephaly is a neurodevelopmental disorder defined by an abnormally small brain, and many gene mutations have been found to be associated with primary microcephaly. However, mouse models generated by ablating primary microcephaly-associated genes often fail to recapitulate the severe loss of cortical surface area observed in individuals with this pathology. Here, we show that a mouse model with deficient expression of high-mobility group nucleosomal binding domain 2 (HMGN2) manifests microcephaly with reduced cortical surface area and almost normal radial corticogenesis, with a pattern of incomplete penetrance. We revealed that altered cleavage plane and mitotic delay of ventricular radial glia may explain the rising ratio of intermediate progenitor cells to radial glia and the displacement of neural progenitor cells in microcephalic mutant mice. These led to decreased self-renewal of the radial glia and reduction in lateral expansion. Furthermore, we found that HMGN2 protected corticogenesis by maintaining global chromatin accessibility mainly at promoter regions, thereby ensuring the correct regulation of the transcriptome. Our findings underscore the importance of the regulation of chromatin structure in cortical development and highlight a mouse model with critical insights into the etiology of microcephaly.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution