Merck
  • Monitoring the glycosylation of α-amino-3-hydroxy-5-methyl-4-isoxazole-propionate-type glutamate receptors using specific antibodies reveals a novel regulatory mechanism of N-glycosylation occupancy by molecular chaperones in mice.
Monitoring the glycosylation of α-amino-3-hydroxy-5-methyl-4-isoxazole-propionate-type glutamate receptors using specific antibodies reveals a novel regulatory mechanism of N-glycosylation occupancy by molecular chaperones in mice.
Journal of neurochemistry (2020-01-21)
Ryosuke Midorikawa, Daisuke Takakura, Jyoji Morise, Yoshihiko Wakazono, Nana Kawasaki, Shogo Oka, Kogo Takamiya
ZUSAMMENFASSUNG
In the mammalian nervous system, protein N-glycosylation plays an important role in neuronal physiology. In this study, we performed a comprehensive N-glycosylation analysis of mouse GluA1, one of the major subunits of α-amino-3-hydroxy-5-methyl-4-isoxazole-propionate type glutamate receptor, which possesses six potential N-glycosylation sites in the N-terminal domain. By mass spectrometry-based analysis, we identified the N-glycoforms and semiquantitatively determined the site-specific N-glycosylation occupancy of GluA1. In addition, only the N401-glycosylation site demonstrated incomplete N-glycosylation occupancy. Therefore, we generated a peptide antibody that specifically detects the N401-glycan-free form to precisely quantify N401-glycosylation occupancy. Using this antibody, we clarified that N401 occupancy varies between cell types and increases in an age-dependent manner in mouse forebrains. To address the regulatory mechanism of N401-glycosylation, binding proteins of GluA1 around the N401 site were screened. HSP70 family proteins, including Bip, were identified as candidates. Bip has been known as a molecular chaperone that plays a key role in protein folding in the ER (endoplasmic reticulum). To examine the involvement of Bip in N401-glycosylation, the effect of Bip over-expression on N401 occupancy was evaluated in HEK293T cells, and the results demonstrated Bip increases the N401 glycan-free form by mediating selective prolongation of its protein half-life. Taken together, we propose that the N401-glycosite of GluA1 receives a unique control of modification, and we also propose a novel N-glycosylation occupancy regulatory mechanism by Bip that might be associated with α-amino-3-hydroxy-5-methyl-4-isoxazole-propionate receptors function in the brain.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fetales Kälberserum, non-USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein