Merck
  • Digestibility of amino acids, energy, and minerals in roasted full-fat soybean and expelled-extruded soybean meal fed to growing pigs without or with multienzyme supplement containing fiber-degrading enzymes, protease, and phytase.
Digestibility of amino acids, energy, and minerals in roasted full-fat soybean and expelled-extruded soybean meal fed to growing pigs without or with multienzyme supplement containing fiber-degrading enzymes, protease, and phytase.
Journal of animal science (2020-05-22)
Elijah G Kiarie, Ilona A Parenteau, Cuilan Zhu, Nelson E Ward, Aaron J Cowieson
ZUSAMMENFASSUNG
Indigestible fiber-protein-phytate complexes reduce the feeding value of soy products. We investigated the effects of multienzyme supplement (MES, Victus) on standardized ileal digestibility (SID) of amino acids (AA) and apparent total tract digestibility (ATTD) of energy and minerals in roasted full-fat soybean (FFSB) seeds and expelled-extruded soybean meal (SBM) fed to growing pigs. The crude protein (CP) was 33.4% and 42.8% dry matter (DM) in FFSB seeds and SBM, respectively and corresponding values for crude fat were 17.4% and 11.8% DM. Semi-purified diets with 50% of either FFSB seeds or SBM as the sole source of AA were prepared without or with MES supplying phytase, protease, xylanase, and β-glucanase at 2,200, 8,300, 400, and 100 U/kg of feed, respectively. Diets had TiO2 as an indigestible marker and the ratio of cornstarch to sucrose and corn oil was identical to calculate DE by the difference method. Eight ileal-cannulated barrows (22.1 ± 0.61 kg) were fed diets in a replicated 4 × 4 Latin square design to give eight replicates per diet. The period lasted for 9 d: 5 d for acclimation, 2 d for fecal, and 2 d for ileal digesta samples. There was no (P > 0.05) interaction between soy type and MES or MES effect on SID of AA; SBM had higher (P < 0.05) SID of CP, His, Leu, and Lys. There was no (P > 0.05) interaction between soy type and MES on energy digestibility. The FFSB seeds had higher ATTD of gross energy (GE, 80.2% vs. 76.6%; P < 0.01) than SBM. Pigs fed MES had higher (P < 0.05) ATTD of DM (91.3% vs. 87.7 %), GE (87.5% vs. 82.4%), CP (86.4% vs. 82.9%), crude fat (70.6% vs. 54.9%), Ca (63.2% vs. 60.2%), and P (67.5% vs. 63.2%). In conclusions, differences on AA and energy digestibility in soy products could be linked to processing and compositional differences. Although MES had no effect on SID of AA, the effects on the utilization of minerals and energy demonstrated the value of fiber-degrading enzymes, protease, and phytase in improving the nutritive value of soy products independent of processing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Amylase, hitzestabil, solution, for use in Total Dietary Fiber Assay, TDF-100A