Merck
  • Long-term observation of airway reconstruction using decellularized tracheal allografts in micro-miniature pigs at growing stage.
Long-term observation of airway reconstruction using decellularized tracheal allografts in micro-miniature pigs at growing stage.
Regenerative therapy (2021-01-12)
Michinobu Ohno, Yasushi Fuchimoto, Masataka Higuchi, Tetsuji Yamaoka, Makoto Komura, Akihiro Umezawa, Huai-Che Hsu, Shin Enosawa, Tatsuo Kuroda
ZUSAMMENFASSUNG
Decellularized tissue exhibits cell matrix-like properties, along with reduced antigenicity. We explored the potential of decellularized allogeneic trachea to restore the upper respiratory tract, focusing on pediatric application. This study specifically aimed at long-term observation of tissue regeneration using a micro-miniature pig model. Artificial defects (15 × 15 mm) in the subglottis and trachea of micro-miniature pigs were repaired by transplantation of either allogeneic decellularized or fresh (control) tracheal patches. Pigs were evaluated in situ, by bronchoscopy, every three months, and sacrificed for histological examination at six and twelve months after transplantation. No airway symptom was observed in any pig during the observation period. Bronchoscopy revealed the tracheal lumen to be restored by fresh grafts, showing an irregular surface with remarkable longitudinal compression; these changes were mild after restoration with decellularized grafts. Histologically, while fresh graft patches were denatured and replaced by calcified tissue, decellularized patches remained unchanged throughout the observation period. There were regeneration foci of cartilage adjacent to the grafts, and some foci joined the decellularized graft uniformly, suggesting the induction of tracheal reconstitution. Allogeneic decellularized tracheal tissue could serve as a promising biomaterial for tracheal restoration, especially for pediatric patients at the growing stage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DNAse I, Amplification Grade