Merck
Nutraceutical Potential of Five Mexican Brown Seaweeds.
BioMed research international (2020-01-14)
Ana Rocío Múzquiz de la Garza, Mireya Tapia-Salazar, Maribel Maldonado-Muñiz, Julián de la Rosa-Millán, Janet Alejandra Gutiérrez-Uribe, Liliana Santos-Zea, Bertha Alicia Barba-Dávila, Denis Ricque-Marie, Lucía Elizabeth Cruz-Suárez
ZUSAMMENFASSUNG
In search of pharmaceutically active products to control type 2 diabetes, five brown seaweeds (Silvetia compressa, Cystoseira osmundacea, Ecklonia arborea, Pterygophora californica, and Egregia menziesii) from the Northwest Mexican Pacific coast were investigated. Proximate composition and total polyphenol content (TPC) as phloroglucinol equivalents (PGE) were determined for the five seaweed powders and their respective hydroethanolic (1 : 1) extracts. Extracts were screened for their radical scavenging activity (DPPH and ORAC) and glycosidase inhibitory activity. HPLC-DAD, HPLC-MS-TOF, and ATR-FT-IR methodologies were used to identify the most abundant phlorotannins and sulfated polysaccharides in the extracts. Hydroethanolic extracts contained minerals (17 to 59% of the dry matter), proteins (4 to 9%), ethanol-insoluble polysaccharides (5.4 to 53%), nitrogen-free extract (NFE) (24.4 to 70.1%), lipids (5 to 12%), and TPC (2.6 to 47.7 g PGE per 100 g dry extract). S. compressa and E. arborea dry extracts presented the lowest ash content (26 and 17%, respectively) and had some of the highest phenolic (47.7 and 15.2 g PGE per 100 g extract), NFE (57.3 and 70.1%), and soluble polysaccharide (19.7 and 53%) contents. S. compressa and E. arborea extracts had the highest antioxidant activity (IC50 DPPH 1.7 and 3.7 mg mL-1; ORAC 0.817 and 0.801 mmol Trolox equivalent/g extract) and the highest α-amylase and α-glucosidase inhibitory capacities (IC50 940 and 1152 μg mL-1 against α-amylase and 194 and 647 μg mL-1 against α-glucosidase). The most abundant phlorotannins identified in the extracts were phloretol, fucophloroethol, and two- and three-phloroglucinol unit (PGU) phlorotannins. Laminarin, fucoidan, and alginate were among the sulfated polysaccharides identified in the extracts. The bioactivities of S. compressa and E. arborea extracts were mainly related with their contents of three PGU phlorotannins and sulfated polysaccharides (e.g., fucoidan, laminarin, and alginate). These results suggest S. compressa and E. arborea are potential candidates for food products and nutraceutical and pharmaceutical preparations, and as additives for diabetes management.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Fluoreszein Natriumsalz, used as fluorescent tracer