- A drug screening toolkit based on the -1 ribosomal frameshifting of SARS-CoV-2.
A drug screening toolkit based on the -1 ribosomal frameshifting of SARS-CoV-2.
Heliyon (2020-09-02)
Yanqiong Chen, Huan Tao, Silan Shen, Zhiyong Miao, Lili Li, Yongqian Jia, Hu Zhang, Xiufeng Bai, Xinyuan Fu
PMID32869005
ZUSAMMENFASSUNG
The -1 ribosomal frameshifting is vital for the translation of the open reading frame (ORF)1b in SARS-CoV-2. The products of ORF1b participate in viral replication. Therefore, changing the frameshift frequency reduces the survival of the virus. This study aimed to successfully develop a toolkit for screening antiviral drugs. Finally, the FDA-approved drug library was screened, revealing that ivacaftor and (-)-Huperzine A worked well in changing the -1 ribosomal frameshifting of SARS-CoV-2 in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Polyethylenimin, verzweigt, average Mw ~25,000 by LS, average Mn ~10,000 by GPC, branched