Merck
  • Increased circulating levels of Factor H-Related Protein 4 are strongly associated with age-related macular degeneration.
Increased circulating levels of Factor H-Related Protein 4 are strongly associated with age-related macular degeneration.
Nature communications (2020-02-09)
Valentina Cipriani, Laura Lorés-Motta, Fan He, Dina Fathalla, Viranga Tilakaratna, Selina McHarg, Nadhim Bayatti, İlhan E Acar, Carel B Hoyng, Sascha Fauser, Anthony T Moore, John R W Yates, Eiko K de Jong, B Paul Morgan, Anneke I den Hollander, Paul N Bishop, Simon J Clark
ZUSAMMENFASSUNG
Age-related macular degeneration (AMD) is a leading cause of blindness. Genetic variants at the chromosome 1q31.3 encompassing the complement factor H (CFH, FH) and CFH related genes (CFHR1-5) are major determinants of AMD susceptibility, but their molecular consequences remain unclear. Here we demonstrate that FHR-4 plays a prominent role in AMD pathogenesis. We show that systemic FHR-4 levels are elevated in AMD (P-value = 7.1 × 10-6), whereas no difference is seen for FH. Furthermore, FHR-4 accumulates in the choriocapillaris, Bruch's membrane and drusen, and can compete with FH/FHL-1 for C3b binding, preventing FI-mediated C3b cleavage. Critically, the protective allele of the strongest AMD-associated CFH locus variant rs10922109 has the highest association with reduced FHR-4 levels (P-value = 2.2 × 10-56), independently of the AMD-protective CFHR1-3 deletion, and even in those individuals that carry the high-risk allele of rs1061170 (Y402H). Our findings identify FHR-4 as a key molecular player contributing to complement dysregulation in AMD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Complement factor H from human plasma, 1 mg/mL in PBS, pH 7.2, ≥90% (SDS-PAGE)