Merck
  • Nano-micelle Curcumin; A Hazardous and/or Boosting Agent? Relation with Oocyte In-vitro Maturation and Pre-implantation Embryo Development in Rats.
Nano-micelle Curcumin; A Hazardous and/or Boosting Agent? Relation with Oocyte In-vitro Maturation and Pre-implantation Embryo Development in Rats.
Iranian journal of pharmaceutical research : IJPR (2020-11-24)
Moona Roshanfekr Rad, Vahid Nejati, Mazdak Razi, Gholamreza Najafi
ZUSAMMENFASSUNG
The present study was done to uncover the possible beneficial and/or detrimental effect(s) of nano-micelle curcumin (NMC) on oocyte in-vitro maturation and pre-implantation embryo development. Forty-eight mature female Wistar rats were assigned to control, 7.5, 15, and 30 mg/kg-1 NMC-receiving (orally, for 48 days) groups. To assess the cumulus-oocyte complexes (COCs), the ovaries were stimulated by administrating (i.p.) a 25 IU of the pregnant mare's serum gonadotropin (PMSG) hormone. Following 48-h, 15 IU of hCG was injected (i.p.), and the COCs were taken after 16-18-h. To analyze the pre-implantation embryo development ratio, the sperms were collected from clinically healthy male Wistar rats, and 3.0-3.6 × 106 per mL was added into the fertilization drop. The animals in 7.5 mg/kg-1 NMC-receiving group exhibited a higher oocyte number versus control and other NMC-receiving groups. The NMC, in a dose-dependent manner, decreased the Zygote, 2-cell, blastocyst percentages, as well as hatched embryos, compared to the control group (P < 0.05). The 15 and 30 mg/kg-1 NMC-receiving groups represented a remarkable enhancement in type I arrest. Meanwhile, a significant (P < 0.05) reduction was revealed in type III embryo arrest in the same groups. The NMC, at 7.5 mg/kg-1 potentially enhances the oocyte number, while it fairly reduces the pre-implantation embryo development, even when it is administrated in dose levels of 7.5 mg/kg-1 and/or higher. Although more studies are needed, the NMC could be considered as a suppressor of fertility potential, when consumed chronically even in low doses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fetales Kälberserum, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma