Merck
  • Rotavirus infection induces glycan availability to promote ileum-specific changes in the microbiome aiding rotavirus virulence.
Rotavirus infection induces glycan availability to promote ileum-specific changes in the microbiome aiding rotavirus virulence.
Gut microbes (2020-05-15)
Melinda A Engevik, Lori D Banks, Kristen A Engevik, Alexandra L Chang-Graham, Jacob L Perry, Diane S Hutchinson, Nadim J Ajami, Joseph F Petrosino, Joseph M Hyser
ZUSAMMENFASSUNG
Multiple studies have identified changes within the gut microbiome in response to diarrheal-inducing bacterial pathogens. However, examination of the microbiome in response to viral pathogens remains understudied. Compounding this, many studies use fecal samples to assess microbiome composition; which may not accurately mirror changes within the small intestine, the primary site for most enteric virus infections. As a result, the functional significance of small intestinal microbiome shifts during infection is not well defined. To address these gaps, rotavirus-infected neonatal mice were examined for changes in bacterial community dynamics, host gene expression, and tissue recovery during infection. Profiling bacterial communities using 16S rRNA sequencing suggested significant and distinct changes in ileal communities in response to rotavirus infection, with no significant changes for other gastrointestinal (GI) compartments. At 1-d post-infection, we observed a loss in Lactobacillus species from the ileum, but an increase in Bacteroides and Akkermansia, both of which exhibit mucin-digesting capabilities. Concomitant with the bacterial community shifts, we observed a loss of mucin-filled goblet cells in the small intestine at d 1, with recovery occurring by d 3. Rotavirus infection of mucin-producing cell lines and human intestinal enteroids (HIEs) stimulated release of stored mucin granules, similar to in vivo findings. In vitro, incubation of mucins with Bacteroides or Akkermansia members resulted in significant glycan degradation, which altered the binding capacity of rotavirus in silico and in vitro. Taken together, these data suggest that the response to and recovery from rotavirus-diarrhea is unique between sub-compartments of the GI tract and may be influenced by mucin-degrading microbes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Formalinlösung, neutral gepuffert, 10 %, histological tissue fixative
Sigma-Aldrich
Fluoromount Wässriges Einbettungsmedium, for use with fluorescent dye-stained tissues
Sigma-Aldrich
Mucin aus Schweinemagen, Type II
Sigma-Aldrich
bisBenzimid H 33342, ≥98% (HPLC and TLC)