Merck
  • In vitro chemosensitization of Plasmodium falciparum to antimalarials by verapamil and probenecid.
In vitro chemosensitization of Plasmodium falciparum to antimalarials by verapamil and probenecid.
Antimicrobial agents and chemotherapy (2009-04-15)
Victor Masseno, Steven Muriithi, Alexis Nzila
ZUSAMMENFASSUNG
We tested the effect of probenecid and verapamil in chemosensitizing Plasmodium falciparum to 14 antimalarials using the multidrug-resistant strain V1S and the drug-sensitive 3D7. Verapamil chemosensitizes V1S to quinine and chloroquine. Interestingly, probenecid profoundly chemosensitizes V1S to piperaquine. Thus, probenecid could be used to increase piperaquine efficacy in vivo.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chinin, 90%
Sigma-Aldrich
Chinin, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)