Merck
  • m6 A demethylase ALKBH5 promotes proliferation of esophageal squamous cell carcinoma associated with poor prognosis.
m6 A demethylase ALKBH5 promotes proliferation of esophageal squamous cell carcinoma associated with poor prognosis.
Genes to cells : devoted to molecular & cellular mechanisms (2020-05-26)
Yushi Nagaki, Satoru Motoyama, Tomokazu Yamaguchi, Midori Hoshizaki, Yusuke Sato, Teruki Sato, Yukio Koizumi, Akiyuki Wakita, Yuta Kawakita, Kazuhiro Imai, Hiroshi Nanjo, Hiroyuki Watanabe, Yumiko Imai, Yoshihiro Minamiya, Keiji Kuba
ZUSAMMENFASSUNG
Esophageal squamous cell carcinoma (ESCC) is one of the most fatal types of malignant tumors worldwide. Epitranscriptome, such as N6 -methyladenosine (m6 A) of mRNA, is an abundant post-transcriptional mRNA modification and has been recently implicated to play roles in several cancers, whereas the significance of m6 A modifications is virtually unknown in ESCC. Analysis of tissue microarray of the tumors in 177 ESCC patients showed that higher expression of m6 A demethylase ALKBH5 correlated with poor prognosis and that ALKBH5 was an independent prognostic factor of the survival of patients. There was no correlation between the other demethylase FTO and prognosis. siRNA knockdown of ALKBH5 but not FTO significantly suppressed proliferation and migration of human ESCC cells. ALKBH5 knockdown delayed progression of cell cycle and accumulated the cells to G0/G1 phase. Mechanistically, expression of CDKN1A (p21) was significantly up-regulated in ALKBH5-depleted cells, and m6 A modification and stability of CDKN1A mRNA were increased by ALKBH5 knockdown. Furthermore, depletion of ALKBH5 substantially suppressed tumor growth of ESCC cells subcutaneously transplanted in BALB/c nude mice. Collectively, we identify ALKBH5 as the first m6 A demethylase that accelerates cell cycle progression and promotes cell proliferation of ESCC cells, which is associated with poor prognosis of ESCC patients.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
RPMI-1640-Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Roche
cOmplete Mini Proteasehemmer-Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Puromycin -dihydrochlorid, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Demecolcin -Lösung, 10 μg/mL in HBSS, ACF Qualified, BioXtra