Merck
  • What is the response profile of deciduous pulp fibroblasts stimulated with E. coli LPS and E. faecalis LTA?
What is the response profile of deciduous pulp fibroblasts stimulated with E. coli LPS and E. faecalis LTA?
BMC immunology (2020-06-24)
Bella Luna Colombini-Ishikiriama, Thiago Jose Dionisio, Thais Francini Garbieri, Rafaela Alves da Silva, Maria Aparecida Andrade Moreira Machado, Sandra Helena Penha de Oliveira, Vanessa Soares Lara, Andrew Seth Greene, Carlos Ferreira Santos
ZUSAMMENFASSUNG
Oral fibroblast immunological responses to bacterial stimuli are well known. However, there are few studies about pulp fibroblasts from deciduous teeth (HDPF) responses, which are important for the treatment of pulp infections in children. The aim of this study was to evaluate expression and production of inflammatory cytokines and chemokines by HDPF when challenged with bacterial antigens normally present in advanced caries lesions. Triplicate HDPF from 4 children (n = 4; 2 boys and 2 girls) were cultured by explant technique and challenged or not with Escherichia coli lipopolysaccharide/1 μg/mL (EcLPS) or Enterococcus faecalis lipoteichoic acid/1 μg/mL (EfLTA) for 6 and 24 h. Most of published studies employed immortalized cells, i.e., without checking possible gender and genetic variables. mRNA expression and protein production were evaluated by RT-qPCR and ELISA MILLIPLEX®, respectively, for Interleukin (IL)-1α, IL-1β, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12, IL-17, Chemokine C-C motif ligand 2/monocyte chemoattractant protein 1 (CCL2/MCP-1), Chemokine C-C motif ligand 3/macrophage inflammatory protein 1-alpha (CCL3/MIP1-α), Chemokine C-C motif ligand 5/ regulated on activation, normal T cell expressed and secreted (CCL5/RANTES), C-X-C motif chemokine 12/ stromal cell-derived factor 1 (CXCL12/SDF-1), Tumor Necrosis Factor-alpha (TNF-α), Interferon-gamma (IFN γ), Vascular Endothelial Growth Factor (VEGF), Colony stimulating factor 1 (CSF-1) and Macrophage colony-stimulating factor (M-CSF). EcLPS increased IL-1α, IL-1β, IL-8, CCL2, CCL5, TNF-α and CSF-1 mRNA and protein levels while EfLTA was only able to positively regulate gene expression and protein production of IL-8. The results of the present study confirmed our hypothesis, since pulp fibroblasts from deciduous teeth are capable of increasing gene expression and protein production after being stimulated with EcLPS and EfLTA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, γ-irradiated, BioXtra, suitable for cell culture