Merck
  • AZD9291 Resistance Reversal Activity of a pH-Sensitive Nanocarrier Dual-Loaded with Chloroquine and FGFR1 Inhibitor in NSCLC.
AZD9291 Resistance Reversal Activity of a pH-Sensitive Nanocarrier Dual-Loaded with Chloroquine and FGFR1 Inhibitor in NSCLC.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2021-01-30)
Yu Gu, Songtao Lai, Yang Dong, Hao Fu, Liwei Song, Tianxiang Chen, Yourong Duan, Zhen Zhang
ZUSAMMENFASSUNG
AZD9291 can effectively prolong survival of non-small cell lung cancer (NSCLC) patients. Unfortunately, the mechanism of its acquired drug resistance is largely unknown. This study shows that autophagy and fibroblast growth factor receptor 1 signaling pathways are both activated in AZD9291 resistant NSCLC, and inhibition of them, respectively, by chloroquine (CQ) and PD173074 can synergistically reverse AZD9291 resistance. Herein, a coloaded CQ and PD173074 pH-sensitive shell-core nanoparticles CP@NP-cRGD is developed to reverse AZD9291 resistance in NSCLC. CP@NP-cRGD has a high encapsulation rate and stability, and can effectively prevent the degradation of drugs in circulation process. CP@NP-cRGD can target tumor cells by enhanced permeability and retention effect and the cRGD peptide. The pH-sensitive CaP shell can realize lysosome escape and then release drugs successively. The combination of CP@NP-cRGD and AZD9291 significantly induces a higher rate of apoptosis, more G0/G1 phase arrest, and reduces proliferation of resistant cell lines by downregulation of p-ERK1/2 in vitro. CQ in CP@NP-cRGD can block protective autophagy induced by both AZD9291 and PD173074. CP@NP-cRGD combined with AZD9291 shows adequate tumor enrichment, low toxicity, and excellent antitumor effect in nude mice. It provides a novel multifunctional nanoparticle to overcome AZD9291 resistance for potential clinical applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chloroquin -diphosphat (Salz), powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Sigma-Aldrich
Rhodamin B, ≥95% (HPLC)
Sigma-Aldrich
Protease- und Phosphatasehemmer-Cocktail
Sigma-Aldrich
Dansylcadaverin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)