Merck
  • New imidazopyridines with phosphodiesterase 4 and 7 inhibitory activity and their efficacy in animal models of inflammatory and autoimmune diseases.
New imidazopyridines with phosphodiesterase 4 and 7 inhibitory activity and their efficacy in animal models of inflammatory and autoimmune diseases.
European journal of medicinal chemistry (2020-10-07)
Veronika Ručilová, Artur Świerczek, David Vanda, Petr Funk, Barbora Lemrová, Alicja Gawalska, Adam Bucki, Barbara Nowak, Monika Zadrożna, Krzysztof Pociecha, Miroslav Soural, Elżbieta Wyska, Maciej Pawłowski, Grażyna Chłoń-Rzepa, Paweł Zajdel
ZUSAMMENFASSUNG
Herein, we describe the rapid synthesis of a focused library of trisubstituted imidazo[4,5-b]pyridines and imidazo[4,5-c]pyridines from 2,4-dichloro-3-nitropyridine using the combination of solution-phase/solid-phase chemistry as new potential anti-inflammatory agents in the treatment of autoimmune diseases. Structure-activity relationship studies, followed by the structure optimization, provided hit compounds (17 and 28) which inhibited phosphodiesterase 4 (PDE4) with IC50 values comparable to rolipram and displayed different inhibitory potency against phosphodiesterase 7 (PDE7). Among them, compound 17 showed a beneficial effect in all the studied animal models of inflammatory and autoimmune diseases (concanavalin A-induced hepatitis, lipopolysaccharide-induced endotoxemia, collagen-induced arthritis, and MOG35-55-induced encephalomyelitis). In addition, compound 17 showed a favorable pharmacokinetic profile after intraperitoneal administration; it was characterized by a fast absorption from the peritoneal cavity and a relatively long terminal half-life in rats. It was found to penetrate brain barrier in mice. The performed experiments sheds light on the impact of PDE7A inhibition for the efficacy of PDE4 inhibitors in these disease conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)