Merck
  • Role of adenomatous polyposis coli in proliferation and differentiation of colon epithelial cells in organoid culture.
Role of adenomatous polyposis coli in proliferation and differentiation of colon epithelial cells in organoid culture.
Scientific reports (2021-02-19)
Daisuke Yamazaki, Osamu Hashizume, Shiho Taniguchi, Yosuke Funato, Hiroaki Miki
ZUSAMMENFASSUNG
Adenomatous polyposis coli (APC) is a tumor-suppressing protein whose inactivation triggers the formation of colorectal polyps. Numerous studies using cell lines or genetically engineered mice have revealed its role in suppressing Wnt/β-catenin signaling pathway and regulating cell proliferation and differentiation. Here, we performed genetic analyses of APC using a three-dimensional organoid culture of mouse colon epithelia, which enables the detailed examination of epithelial properties. Analyses of Apc-knockout colon organoids not only confirmed the importance of APC in suppressing Wnt/β-catenin signaling and regulating cell differentiation, but also revealed several novel features: a significant decrease in proliferating speed and an increase in cross-sectional area of cells. Moreover, we found a significant number of lysozyme-positive Paneth-like cells, which were never observed in wild-type colon tissues or organoids, but have been reported to emerge in colon cancers. Therefore, APC autonomously suppresses ectopic differentiation into lysozyme-positive cells, specifically in the colon epithelia. Colon organoids would be an ideal material to investigate the molecular mechanism and biological importance of the ectopic differentiation associated with cancer development.