Merck
  • Functional reconstruction of human AML reveals stem cell origin and vulnerability of treatment-resistant MLL-rearranged leukemia.
Functional reconstruction of human AML reveals stem cell origin and vulnerability of treatment-resistant MLL-rearranged leukemia.
Science translational medicine (2021-02-26)
Bernd B Zeisig, Tsz Kan Fung, Magdalena Zarowiecki, Chiou Tsun Tsai, Huacheng Luo, Boban Stanojevic, Claire Lynn, Anskar Y H Leung, Jan Zuna, Marketa Zaliova, Martin Bornhauser, Malte von Bonin, Boris Lenhard, Suming Huang, Ghulam J Mufti, Chi Wai Eric So
ZUSAMMENFASSUNG
Chemoresistance remains the major challenge for successful treatment of acute myeloid leukemia (AML). Although recent mouse studies suggest that treatment response of genetically and immunophenotypically indistinguishable AML can be influenced by their different cells of origin, corresponding evidence in human disease is still largely lacking. By combining prospective disease modeling using highly purified human hematopoietic stem or progenitor cells with retrospective deconvolution study of leukemia stem cells (LSCs) from primary patient samples, we identified human hematopoietic stem cells (HSCs) and common myeloid progenitors (CMPs) as two distinctive origins of human AML driven by Mixed Lineage Leukemia (MLL) gene fusions (MLL-AML). Despite LSCs from either MLL-rearranged HSCs or MLL-rearranged CMPs having a mature CD34-/lo/CD38+ immunophenotype in both a humanized mouse model and primary patient samples, the resulting AML cells exhibited contrasting responses to chemotherapy. HSC-derived MLL-AML was highly resistant to chemotherapy and expressed elevated amounts of the multispecific anion transporter ABCC3. Inhibition of ABCC3 by shRNA-mediated knockdown or with small-molecule inhibitor fidaxomicin, currently used for diarrhea associated with Clostridium difficile infection, effectively resensitized HSC-derived MLL-AML toward standard chemotherapeutic drugs. This study not only functionally established two distinctive origins of human LSCs for MLL-AML and their role in mediating chemoresistance but also identified a potential therapeutic avenue for stem cell-associated treatment resistance by repurposing a well-tolerated antidiarrhea drug already used in the clinic.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC3