Merck
  • Inhibition of Amyloid Fibrillation of HEWL by 4-Methylcoumarin and 4-Methylthiocoumarin Derivatives.
Inhibition of Amyloid Fibrillation of HEWL by 4-Methylcoumarin and 4-Methylthiocoumarin Derivatives.
Current pharmaceutical biotechnology (2020-09-17)
Shivani Kumar, Manoj Kumar, Yogesh K Tyagi, Suresh Kumar
ZUSAMMENFASSUNG
Several human diseases like Parkinson's, Alzheimer's disease, and systemic amyloidosis are associated with the misfolding and aggregation of protein molecules. The present study demonstrated the comparison of 4-methyl coumarin and 4-methylthiocoumarin derivative for their anti-amyloidogenic and disaggregation activities. The hen egg-white lysozyme is used as a model system to study protein aggregation and disaggregation under in vitro conditions. Techniques used in the study were Thioflavin T fluorescence assay, intrinsic fluorescence assay, circular dichroism, transmission electron microscopy, and molecular dynamics. Fifteen compounds were screened for their anti-amyloidogenic and disaggregation potential. Six compounds significantly inhibited the fibril formation, whereas ten compounds showed disaggregation property of pre-formed fibrils. Under in vitro conditions, the compound C3 and C7 showed significant inhibition of fibril formation in a concentration-dependent manner as compared to control. C3 and C7 demonstrated 93% and 76% inhibition of fibril formation, respectively. Furthermore, C3 and C7 exhibited 83% and 76% disaggregation activity, respectively, of pre-formed HEWL fibrils at their highest concentration. These anti-amyloidogenic and disaggregation potential of C3 and C7 were validated by intrinsic fluorescence, CD, molecular dynamics, and TEM study. 4-methylthiocoumarins derivatives have shown better anti-amyloidogenic activity as compared to 4-methylcoumarin derivatives for both amyloid formation as well as disaggregation of preformed amyloid fibrils. Structurally, the derivatives of 4-methylthiocoumarins (C3 and C7) contain thio group on 2nd position that might be responsible for anti-amyloidogenic activity as compared to 4- methylcoumarin derivatives (C2 and C4). C3 and C7 are novel 4-methylthiocoumarin derivatives that can be used as a lead for alleviation and symptoms associated with protein aggregation disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioflavin T, used as stain for amyloid