Merck
  • Achieving Purely-Organic Room-Temperature Aqueous Phosphorescence via a Two-Component Macromolecular Self-Assembly Strategy.
Achieving Purely-Organic Room-Temperature Aqueous Phosphorescence via a Two-Component Macromolecular Self-Assembly Strategy.
Chemistry, an Asian journal (2020-09-11)
Wang Guo, Xuepu Wang, Bei Zhou, Kaka Zhang
ZUSAMMENFASSUNG
Manipulation of supramolecular behaviors and aggregation states represents an important topic in devising intriguing photofunctional systems. Here we report a two-component macromolecular self-assembly strategy for achieving aqueous room-temperature phosphorescence (RTP) in purely organic systems. Amphiphilic triblock copolymers are used to modulate the self-assembly of planar RTP molecules in aqueous solution, leading to the formation of sheet-like RTP objects with well-defined morphology, uniform crystalline nanostructures and excellent aqueous dispersity. In contrast, the addition of the planar RTP molecules into aqueous medium only leads to precipitation and quenching of RTP properties. Powder X-ray diffraction and single-crystal X-ray diffraction studies reveal that the amphiphilic triblock copolymers can assist supramolecular columnar packing of the planar RTP molecules where multiple non-covalent interactions stabilize the triplet excited states. Interestingly, it is found that luminescent signals of the sheet-like RTP objects can be extracted from strong fluorescent environments by phosphorescence mode and emission lifetime measurement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Latex-Beads, carboxylatmodifiziertes Polystyrol, gelb-grün fluoreszierend, aqueous suspension, 2.0 μm mean particle size