Merck
  • Oral deoxynivalenol toxicity in Harlan Sprague Dawley (Hsd:Sprague Dawley® SD®) rat dams and their offspring.
Oral deoxynivalenol toxicity in Harlan Sprague Dawley (Hsd:Sprague Dawley® SD®) rat dams and their offspring.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2021-01-04)
Madelyn C Huang, Johnathan R Furr, Veronica G Robinson, Laura Betz, Keith Shockley, Helen Cunny, Kristine Witt, Suramya Waidyanatha, Dori Germolec
ZUSAMMENFASSUNG
There is widespread human exposure to deoxynivalenol (DON), a fungal mycotoxin found globally in many grain-based foods and animal feed. Acute exposures to high levels of DON are associated with gastrointestinal effects and emesis in humans and some animals, but the effects of low-dose exposures throughout the lifetime, a more likely exposure scenario in humans, are understudied. Therefore, this study was designed to identify doses of DON that could be used to evaluate long-term toxicity following perinatal exposure. Time-mated Harlan Sprague Dawley (Hsd:Sprague Dawley® SD®) rats were administered 0, 0.03, 0.1, 0.3, 1, or 3 mg/kg/day of DON once daily via gavage starting on gestational day 6 through postnatal day (PND) 27. F1 animals were administered the same dose as their respective dams via gavage starting on PND 12 until PND 27. Animals were euthanized on PND 28. DON had no effect on maternal body weight or feed consumption at any dose. Findings were limited to the 3 mg/kg/day group: F0 females had smaller live litter sizes than controls and F1 pups had lower body weight (4-13%) compared to controls. By PND 28, F1 body weight, after adjustments for litter effects, was 10-13% lower than controls. Blood samples obtained on PND 28 showed no increases in frequencies of micronucleated immature erythrocytes in either F0 or F1 animals. In summary, doses of DON up to 3 mg/kg/day did not affect maternal survival or body weight. Doses of 3 mg/kg/day resulted in slight toxicity manifested as decreased body weight in the offspring. The no-observed effect level was 1 mg/kg/day.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Diazo-5-oxo-L-Norleucin, powder