Merck
  • Development of selective preconcentration/clean-up method for imidazolinone herbicides determination in natural water and rice samples by HPLC-PAD using an imazethapyr imprinted poly(vinylimidazole-TRIM).
Development of selective preconcentration/clean-up method for imidazolinone herbicides determination in natural water and rice samples by HPLC-PAD using an imazethapyr imprinted poly(vinylimidazole-TRIM).
Food chemistry (2020-07-28)
César Ricardo Teixeira Tarley, Fabio Antonio Cajamarca Suquila, Juliana Casarin, Affonso Celso Gonçalves Junior, Mariana Gava Segatelli
ZUSAMMENFASSUNG
The development of a novel molecularly imprinted solid-phase extraction (MISPE) method for simultaneous preconcentration of imazapyr (IMP), imazapic (IMZ) and imazethapyr (IMT) with determination by HPLC-PAD (High performance liquid chromatography - photodiode-array detector) is proposed. The polymer synthesis was performed using imazethapyr as template molecule and 1-vinylimidazole as functional monomer. The method is based on preconcentration of 100.0 mL of sample through 200.0 mg of molecularly imprinted poly(vinylimidazole-TRIM) (MIP-1VN) at pH 4.0, followed by elution with 2.0 mL of MeOH:CH2Cl2:HAc (34:62:4, v/v). The range of analytical curve (0.29-200.0, 0.21-200.0 and 0.15-200.0 µg L-1), limits of detection (0.09, 0.06 and 0.04 µg L-1) and preconcentration factors (92, 96 and 98) determined for the herbicides, IMP, IMZ and IMT, respectively, were greatly superior when compared with those ones obtained with commercial adsorbents. The analytical method was successfully applied to spiked surface water and rice samples with good results of recovery values (86-107%).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1-Vinylimidazol, ≥99%
Sigma-Aldrich
Trimethylolpropantrimethacrylat, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade
Supelco
Amberlite XAD4, 20-60 mesh
Supelco
Imazapyr, PESTANAL®, analytical standard
Supelco
Imazethapyr, PESTANAL®, analytical standard
Supelco
Imazapic, PESTANAL®, analytical standard
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole