Merck
  • Synthesis, antitubulin, and antiproliferative SAR of analogues of 2-methoxyestradiol-3,17-O,O-bis-sulfamate.
Synthesis, antitubulin, and antiproliferative SAR of analogues of 2-methoxyestradiol-3,17-O,O-bis-sulfamate.
Journal of medicinal chemistry (2010-03-17)
Fabrice Jourdan, Mathew P Leese, Wolfgang Dohle, Ernest Hamel, Eric Ferrandis, Simon P Newman, Atul Purohit, Michael J Reed, Barry V L Potter
ZUSAMMENFASSUNG
The synthesis and antiproliferative activity of analogues of estradiol 3,17-O,O-bis-sulfamates (E2bisMATEs) are discussed. Modifications of the C-17 substituent reveal that an H-bond acceptor is essential for high antiproliferative activity. The local environment in which this H-bond acceptor lies can be varied to an extent. The C-17-oxygen linker can be deleted or substituted with an electronically neutral methylene group, and replacement of the terminal NH(2) with a methyl group is also acceptable. Mesylates 10 and 14 prove equipotent to the E2bisMATEs 2 and 3, while sulfones 20 and 35 display enhanced in vitro antiproliferative activity. In addition, the SAR of 2-substituted estradiol-3-O-sulfamate derivatives as inhibitors of tubulin polymerization has been established for the first time. These agents inhibit the binding of radiolabeled colchicine to tubulin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Methoxyestradiol, powder