Merck
  • Microbial community response to the toxic effect of pentachlorophenol in paddy soil amended with an electron donor and shuttle.
Microbial community response to the toxic effect of pentachlorophenol in paddy soil amended with an electron donor and shuttle.
Ecotoxicology and environmental safety (2020-09-21)
Manjia Chen, Hui Tong, Jiangtao Qiao, Yahui Lv, Qi Jiang, Yuanxue Gao, Chengshuai Liu
ZUSAMMENFASSUNG
Understanding the degradation of pentachlorophenol (PCP) by indigenous microorganisms stimulated by an electron donor and shuttle in paddy soil, and the influences of PCP/electron donor/shuttle on the native microbial community are important for biodegradation and ecological and environmental safety. Previous studies focused on the kinetics and the microbial actions of PCP degradation, however, the effects of toxic and antimicrobial PCP and electron donor/shuttle on the microbial community diversity and composition in paddy soil are poorly understood. In this study, the effects of PCP, an electron donor (lactate), and the electron shuttle (anthraquinone-2, 6-disulfonate, AQDS) on the microbial community in paddy soil were investigated. The results showed that the presence of PCP reduced the microbial diversity compared to the control during PCP degradation, while increased the microbial diversity was observed in response to lactate and AQDS. The addition of PCP stimulated the microorganisms involved in PCP dechlorination, including Clostridium, Desulfitobacterium, Pandoraea, and unclassified Veillonellaceae, which were dormant in raw soil without PCP stress. In all of the treatments with PCP, the addition of lactate or AQDS enhanced PCP dechlorination by stimulating the growth of functional groups involved in PCP dechlorination and by changing the microbial community during dechlorination process. The microbial community tended to be uniform after complete PCP degradation (28 days). However, when lactate and AQDS were present simultaneously in PCP-contaminated soil, lactate acted as a carbon source or electron donor to promote the activities of microbial community, and AQDS changed the redox potential because of the production of reduced AQDS. These findings enhance our understanding of the effect of PCP and a biostimulation method for PCP biodegradation in soil ecosystems at the microbial community level, and suggest the appropriate selection of an electron donor/shuttle for accelerating the bioremediation of PCP-contaminated soils.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Chlor-DL-Phenylalanin
Supelco
2,3,4,5-Tetrachlorphenol, analytical standard